Få mere at vide
Triamcinolone acetonide, MedChemExpress
Artikelnummer. 16481028
Encompass_Preferred
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16481028 5 g
16472249 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16481028 Leverandør Medchem Express Leverandørnr. HYB0636
MedChemExpress Triamcinolone acetonide is a more potent type of triamcinolone, being about 8 times as effective as prednisone.
Triamcinolone acetonide is a more potent type of triamcinolone, being about 8 times as effective as prednisone.
Tekniske data
Kemisk navn eller materiale Triamcinolone acetonide
CAS 76-25-5
Farve White
Renhedsgrad noter Research
Molekylær formel C24H31FO6
Mængde 5 g
Opløselighedsinformation DMSO : 50 mg/mL (115.07 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.23 mM; Need ultrasonic)
SMIL OCC([C@]12[C@@]3([C@@]([C@@]4([H])[C@]([C@H](C3)O)([C@@]5(C(CC4)=CC(C=C5)=O)C)F)([H])C[C@@]1([H])OC(C)(O2)C)C)=O
Molekylvægt (g/mol) 434.5
Formel vægt 434.5
Procent renhed 99.4%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation