Få mere at vide
Trazodone 1,4-Di-N-Oxide, TRC
Artikelnummer. 30455540
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455540 5 mg
30450915 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30455540 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7185155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346603-99-3
Molekylær formel C19H22ClN5O3
Molekylvægt (g/mol) 403.86
Synonym 2-[3-[4-(3-Chlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one 1,4-Dioxide
SMIL ClC1=CC(N2(CCN(CC2)(CCCN3N=C(C=CC=C4)N4C3=O)=O)=O)=CC=C1
Tekniske data
Kemisk navn eller materiale Trazodone 1,4-Di-N-Oxide
Mængde 5 mg
InChI formel InChI=1S/C19H22ClN5O3/c20-16-5-3-6-17(15-16)25(28)13-11-24(27,12-14-25)10-4-9-23-19(26)22-8-2-1-7-18(22)21-23/h1-3,5-8,15H,4,9-14H2
Formel vægt 403.1411