Learn More
VITLAB™ Transparent LDPE Narrow Mouth Bottles
Brand: VITLAB™ 95189
Additional Details : Weight : 0.10000kg
Specifications
Bottle | |
Screw Cap | |
1000 mL | |
LDPE |
215 mm | |
92 mm | |
Screw cap and high shoulder | |
Autoclavable |