missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Transparent LDPE Narrow Mouth Bottles

Brand:  VITLAB™ 138893

Additional Details : Weight : 0.15300kg

 View more versions of this product

Product Code. 10278673

  • 1485.00 DKK / Pack of 25
Estimated Shipment: 09-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Bottle
Screw Cap
2000 mL
LDPE
264 mm
117 mm
Screw cap and flat shoulder
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers