Få mere at vide
trans-Clopidogrel-MP Derivative (Mixture of Diastereomers), TRC
Artikelnummer. 30133109
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30133109 5 mg -
17917444 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133109 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5872555MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1287430-40-3
Molekylær formel C25 H26 Cl N O6 S
Molekylvægt (g/mol) 504
Synonym Clopidogrel Metabolite II, 3-(carboxymethylene)-alpha-(2-chlorophenyl)-4-[[2-(3-methoxyphenyl)-2-oxoethyl]thio]-1-Piperidineacetic acid 1-methyl ester
IUPAC navn (2E)-2-[1-[1-(2-chlorophenyl)-2-methoxy-2-oxoethyl]-4-[2-(3-methoxyphenyl)-2-oxoethyl]sulfanylpiperidin-3-ylidene]acetic acid
SMIL ClC1=CC=CC=C1C(C(OC)=O)N2CCC(SCC(C3=CC(OC)=CC=C3)=O)/C(C2)=C/C(O)=O
Tekniske data
Kemisk navn eller materiale 3-(carboxymethylene)-alpha-(2-chlorophenyl)-4-[(2-(3-methoxyphenyl)-2-oxoethyl)thio]-1-Piperidineacetic acid 1-methyl ester
Mængde 5 mg
InChI formel InChI=1S/C25H26ClNO6S/c1-32-18-7-5-6-16(12-18)21(28)15-34-22-10-11-27(14-17(22)13-23(29)30)24(25(31)33-2)19-8-3-4-9-20(19)26/h3-9,12-13,22,24H,10-11,14-15H2,1-2H3,(H,29,30)/b17-13+
Formel vægt 503.1169