Få mere at vide
trans-9,10-Dihydroxy-9,10-dihydrophenanthrene, TRC
Artikelnummer. 30136103
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136103 500 mg
30011553 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136103 Leverandør Toronto Research Chemicals Leverandørnr. TRCD452755500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 572-41-8
Molekylær formel C14H12O2
Molekylvægt (g/mol) 212.24
Synonym (9R,10R)-rel-9,10-dihydro-9,10-Phenanthrenedioll, trans-9,10-Dihydro-9,10-Phenanthrenediol, (±)-trans-9,10-Dihydro-9,10-dihydroxyphenanthrene;
SMIL O[C@H]1[C@H](O)c2ccccc2c3ccccc13
Tekniske data
Kemisk navn eller materiale trans-9,10-Dihydroxy-9,10-dihydrophenanthrene
Mængde 500 mg
InChI formel InChI=1S/C14H12O2/c15-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14(13)16/h1-8,13-16H/t13-,14-/m0/s1
Formel vægt 212.0837