Learn More
trans-5-Decen-1-ol, 96%
CAS: 56578-18-8 | C10H20O | 156.27 g/mol
563.12 DKK - 1885.00 DKK
Chemical Identifiers
CAS | 56578-18-8 |
---|---|
Molecular Formula | C10H20O |
Molecular Weight (g/mol) | 156.27 |
MDL Number | MFCD00056187 |
InChI Key | WYPQHXVMNVEVEB-AATRIKPKSA-N |
Synonym | trans-5-decen-1-ol, e-5-decen-1-ol, trans-5-decenol, 5-decen-1-ol, 5e, 5-decen-1-ol, e, decan-5-en-1-ol, e-5-decenol, unii-m709v74ozv, 5-decen-1-ol, epa pesticide chemical code 078038 |
PubChem CID | 5283292 |
IUPAC Name | (E)-dec-5-en-1-ol |
SMILES | CCCC\C=C\CCCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11302617
|
Thermo Scientific Alfa Aesar
L11768.03 |
1 g | N/A | ||||||
11312617
|
Thermo Scientific Alfa Aesar
L11768.06 |
5 g |
1885.00 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56578-18-8 | |
156.27 | |
WYPQHXVMNVEVEB-AATRIKPKSA-N | |
5283292 | |
CCCC\C=C\CCCCO |
C10H20O | |
MFCD00056187 | |
trans-5-decen-1-ol, e-5-decen-1-ol, trans-5-decenol, 5-decen-1-ol, 5e, 5-decen-1-ol, e, decan-5-en-1-ol, e-5-decenol, unii-m709v74ozv, 5-decen-1-ol, epa pesticide chemical code 078038 | |
(E)-dec-5-en-1-ol |
Specifications
56578-18-8 | |
62°C (143°F) | |
1.452 | |
1840630 | |
WYPQHXVMNVEVEB-AATRIKPKSA-N | |
(E)-dec-5-en-1-ol | |
5283292 | |
96% |
113°C to 118°C (11 mmHg) | |
C10H20O | |
MFCD00056187 | |
trans-5-decen-1-ol, e-5-decen-1-ol, trans-5-decenol, 5-decen-1-ol, 5e, 5-decen-1-ol, e, decan-5-en-1-ol, e-5-decenol, unii-m709v74ozv, 5-decen-1-ol, epa pesticide chemical code 078038 | |
CCCC\C=C\CCCCO | |
156.27 | |
156.27 | |
trans-5-Decen-1-ol |
Safety and Handling
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 260-267-6
missing translation for 'rtecsNumber' : HE2096000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.