missing translation for 'onlineSavingsMsg'
Learn More
trans-5-Decen-1-ol, 96% Product Code.: 11302617

trans-5-Decen-1-ol, 96%

Product Code. 11302617
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11302617

Brand: Thermo Scientific Alfa Aesar L11768.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 56578-18-8 | C10H20O | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56578-18-8
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00056187
InChI Key WYPQHXVMNVEVEB-AATRIKPKSA-N
Synonym trans-5-decen-1-ol, e-5-decen-1-ol, trans-5-decenol, 5-decen-1-ol, 5e, 5-decen-1-ol, e, decan-5-en-1-ol, e-5-decenol, unii-m709v74ozv, 5-decen-1-ol, epa pesticide chemical code 078038
PubChem CID 5283292
IUPAC Name (E)-dec-5-en-1-ol
SMILES CCCC\C=C\CCCCO

Specifications

Boiling Point 113°C to 118°C (11 mmHg)
Flash Point 62°C (143°F)
Refractive Index 1.452
Quantity 1 g
Beilstein 1840630
Formula Weight 156.27
Percent Purity 96%
Chemical Name or Material trans-5-Decen-1-ol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number10196855 Certificate TypeCertificate of Analysis Date06/07/2023 Product CodeL11768,L11768.03,L11768.06
Lot Number10196855 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeL11768.03,L11768.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-5-Decen-1-ol, 96% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.