Få mere at vide
trans-5,6-Dihydrodihydroxynaphthol, TRC
Artikelnummer. 30482618
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30482618 10 mg -
30482763 2.5 mg 2.5mg
30477395 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30482618 Leverandør Toronto Research Chemicals Leverandørnr. TRCN54265510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38028-40-9
Molekylær formel C10H10O3
Molekylvægt (g/mol) 178.18
Synonym trans-1,2-Dihydro-1,2,5-trihydroxynaphthalin, 1,2-Dihydro-1,2,5-naphthalenetriol
IUPAC navn (1S,2S)-1,2-dihydronaphthalene-1,2,5-triol
SMIL O[C@H]1C=Cc2c(O)cccc2[C@@H]1O
Tekniske data
Kemisk navn eller materiale trans-5,6-Dihydrodihydroxynaphthol
Mængde 10 mg
InChI formel InChI=1S/C10H10O3/c11-8-3-1-2-7-6(8)4-5-9(12)10(7)13/h1-5,9-13H/t9-,10-/m0/s1
Formel vægt 178.063