missing translation for 'onlineSavingsMsg'
Få mere at vide

trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl, 97%, Thermo Scientific™

Artikelnummer. 15462266
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
15462266 5 g 5g
15452266 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15462266 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H55748.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
trans-4-n-Propyl-4′-(3,4,5-trifluorophenyl)bicyclohexyl is used to make PDLC mixtures and Switchable Film . It is also mainly used as a liquid crystal monomer, liquid crystal material.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 131819-23-3
Molekylær formel C21H29F3
Molekylvægt (g/mol) 338.458
MDL nummer MFCD09838998
InChI nøgle FEWMLRARKGRCCE-UHFFFAOYSA-N
Synonym trans,trans-4'-propyl-4-3,4,5-trifluorophenyl bicyclohexyl, trans-4-3,4,5-trifluorophenyl-trans-4'-propylbicyclohexane, trans-4-n-propyl-4'-3,4,5-trifluorophenyl bicyclohexyl, benzene, 1,2,3-trifluoro-5-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4,5-trifluorophenyl-4'-propyl-1,1'-bicyclohexane, trans,trans-1-4'-propylbicyclohexyl-3,4,5-trifluorobenzene, 1,2,3-trifluoro-5-trans,trans-4-propyl 1,1-bicyclohexyl-4-yl benzene, all-trans-4-propyl-4'-3,4,5-trifluorophenyl bi cyclohexane, 1s,1'r,4r,4'r-4-propyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, 1s,4r-4-propyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane
PubChem CID 11772113
IUPAC navn 1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)cyclohexyl]benzene
SMIL CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F

Tekniske data

Kemisk navn eller materiale trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl
Smeltepunkt 63°C to 64°C
Mængde 5 g
Opløselighedsinformation It is soluble in water.
Formel vægt 338.46
Procent renhed 97%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.