Få mere at vide
Thermo Scientific™ Toremifene, 98+%
A chlorinated tamoxifen analogue that induces apoptosis in some cells
Brand: Thermo Scientific™ J63803.03
| Mængde | 1 g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| 89778-26-7 | |
| Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine | |
| A chlorinated tamoxifen analogue that induces apoptosis in some cells | |
| 108°C to 110°C | |
| ≥98% |
| 405.96g/mol | |
| Powder | |
| White to Off-white | |
| Apoptosis Reagent |
Tekniske data
| White to Off-white | |
| Powder | |
| C26H28ClNO | |
| Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine | |
| Apoptosis Reagent | |
| ≥98% |
| 1 g | |
| 89778-26-7 | |
| 405.96g/mol | |
| 108°C to 110°C | |
| A chlorinated tamoxifen analogue that induces apoptosis in some cells |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.