Få mere at vide
Tonabersat, MedChemExpress
Artikelnummer. 16482477
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16482477 50 mg
16446368 10 mg
16428957 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16482477 Leverandør Medchem Express Leverandørnr. HY15204
MedChemExpress Tonabersat (SB-220453) is a gap-junction modulator. Tonabersat prevents inflammatory damage in the central nervous system.
Tonabersat (SB-220453) is a gap-junction modulator. Tonabersat prevents inflammatory damage in the central nervous system.
Tekniske data
Kemisk navn eller materiale Tonabersat
CAS 175013-84-0
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C20H19ClFNO4
Mængde 50 mg
Synonym SB-220453
Opløselighedsinformation DMSO : ≥ 100 mg/mL (255.22 mM)
SMIL O[C@@H]1C(C)(C)OC2=CC=C(C(C)=O)C=C2[C@@H]1NC(C3=CC(Cl)=C(F)C=C3)=O
Molekylvægt (g/mol) 391.82
Formel vægt 391.82
Procent renhed 98.01%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation