Få mere at vide
Tomeglovir, MedChemExpress
Artikelnummer. 16492976
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16492976 25 mg
16490527 50 mg
16484178 10 mg
16476887 5 mg
16405146 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16492976 Leverandør Medchem Express Leverandørnr. HY108261
MedChemExpress Tomeglovir is a potent anti-CMV agent, inhibiting processing of viral DNA-concatemers, with IC50s of 0.34 μM and 0.039 μM for HCMV and MCMV.
Tomeglovir is a potent anti-CMV agent, inhibiting processing of viral DNA-concatemers, with IC50s of 0.34 μM and 0.039 μM for HCMV and MCMV.
Tekniske data
Kemisk navn eller materiale Tomeglovir
CAS 233254-24-5
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C23H27N3O4S
Mængde 25 mg
Synonym BAY 38-4766
Opløselighedsinformation DMSO : ≥ 108 mg/mL (244.60 mM)
SMIL CC(C)(CO)C(NC1=CC=C(NS(=O)(C2=C3C=CC=C(N(C)C)C3=CC=C2)=O)C=C1)=O
Molekylvægt (g/mol) 441.54
Formel vægt 441.54
Procent renhed 99.45%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-anti-virus