Learn More
Tofogliflozin (hydrate), MedChemExpress
MedChemExpress Tofogliflozin hydrate (CSG-452 hydrate) is a potent and highly specific sodium/glucose cotransporter 2 (SGLT2) inhibitor with an IC50 of 2.9 nM and Ki values of 2.9 nM, 14.9 nM, and 6.4 nM for human, rat, and mouse SGLT2. Tofogliflozin partially inhibits high glucose-induced reactive oxyen species (ROS) generation in tubular cells.

1148.12 DKK - 6183.73 DKK
Specifications
Chemical Name or Material | Tofogliflozin (hydrate) |
---|---|
CAS | 1201913-82-7 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C22H28O7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16485668
![]() |
Medchem Express
HY-13413 |
10 mg |
1832.96 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16438367
![]() |
Medchem Express
HY-13413 |
5 mg |
1148.12 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16431837
![]() |
Medchem Express
HY-13413 |
50 mg |
6183.73 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tofogliflozin hydrate (CSG-452 hydrate) is a potent and highly specific sodium/glucose cotransporter 2 (SGLT2) inhibitor with an IC50 of 2.9 nM and Ki values of 2.9 nM, 14.9 nM, and 6.4 nM for human, rat, and mouse SGLT2. Tofogliflozin partially inhibits high glucose-induced reactive oxyen species (ROS) generation in tubular cells.Specifications
Tofogliflozin (hydrate) | |
White | |
C22H28O7 | |
DMSO : ≥ 100 mg/mL (247.25 mM) ∣H2O : 0.33 mg/mL (0.82 mM; Need ultrasonic) | |
404.45 | |
98.73% | |
Solid |
1201913-82-7 | |
Research | |
CSG-452 hydrate | |
O[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@]21OCC3=C2C=C(CC4=CC=C(CC)C=C4)C=C3.O | |
404.45 | |
Research | |
Metabolism-sugar/lipid metabolism |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.