Få mere at vide
Tofacitinib Impurity P, TRC
Artikelnummer. 30456494
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456494 100 mg
30453139 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30456494 Leverandør Toronto Research Chemicals Leverandørnr. TRCT528025100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1252883-90-1
Molekylær formel C20H25N5
Molekylvægt (g/mol) 335.45
Synonym N-Methyl-N-[(3S,4S)-4-methyl-1-(phenylmethyl)-3-piperidinyl]-7H-pyrrolo[2,3-d]pyrimidin-4-amine, N-((3S,4S)-1-Benzyl-4-methylpiperidin-3-yl)-N-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine
IUPAC navn N-[(3S,4S)-1-benzyl-4-methylpiperidin-3-yl]-N-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine
SMIL C[C@H]1CCN(Cc2ccccc2)C[C@H]1N(C)c3ncnc4[nH]ccc34
Tekniske data
Kemisk navn eller materiale Tofacitinib Impurity P
Mængde 100 mg
InChI formel InChI=1S/C20H25N5/c1-15-9-11-25(12-16-6-4-3-5-7-16)13-18(15)24(2)20-17-8-10-21-19(17)22-14-23-20/h3-8,10,14-15,18H,9,11-13H2,1-2H3,(H,21,22,23)/t15-,18+/m0/s1
Formel vægt 335.211