Learn More
Tirapazamine, MedChemExpress
MedChemExpress Tirapazamine is an anticancer agent that shows selective cytotoxicity for hypoxic cells in solid tumors, thereby inducing single-and double-strand breaks in DNA, base damage, and cell death. Tirapazamine is an anticancer and bioreductive agent. Tirapazamine can enhance the cytotoxic effects of ionizing radiation in hypoxic cells.

825.84 DKK - 2749.45 DKK
Specifications
Chemical Name or Material | Tirapazamine |
---|---|
Physical Form | Solid |
CAS | 27314-97-2 |
Color | Orange Red |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16476316
![]() |
Medchem Express
HY-13767 |
100 mg |
1510.69 DKK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16431997
![]() |
Medchem Express
HY-13767 |
50 mg |
825.84 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16431106
![]() |
Medchem Express
HY-13767 |
200 mg |
2749.45 DKK
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tirapazamine is an anticancer agent that shows selective cytotoxicity for hypoxic cells in solid tumors, thereby inducing single-and double-strand breaks in DNA, base damage, and cell death. Tirapazamine is an anticancer and bioreductive agent.Specifications
Tirapazamine | |
27314-97-2 | |
Research | |
Cancer-programmed cell death | |
DMSO : 62.5 mg/mL (350.83 mM; Need ultrasonic) | |
178.15 | |
97.59% |
Solid | |
Orange Red | |
C7H6N4O2 | |
SR259075 SR4233 Win59075 SML 0552 SR 259075 Tirazone | |
NC1=[N+]([O-])C2=CC=CC=C2[N+]([O-])=N1 | |
178.15 | |
Research |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.