Learn More
Lovibond™ Tintometer™ DPD 1 and DPD 3 High Calcium Tablets Combi-Pack
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
369.00 DKK - 770.00 DKK
Specifications
For Use With (Application) | Photometry Tablet Reagents |
---|---|
For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
Type | DPD 1 and DPD 3 High Calcium Tablets Combi-Pack |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15381975
|
Lovibond™
517782BT |
500 Tablets |
770.00 DKK
Pack of 250 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
15371975
|
Lovibond™
517781BT |
250 Tablets |
369.00 DKK
Pack of 100 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
DPD 1 and DPD 3 High Calcium Tablets Combi-Pack |