missing translation for 'onlineSavingsMsg'
Få mere at vide

Timapiprant, MedChemExpress

Artikelnummer. 16442567
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16442567 50 mg
16419057 5 mg
16406468 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16442567 Leverandør Medchem Express Leverandørnr. HY15342

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Timapiprant (OC000459) is a potent, selective, and orally active D prostanoid receptor 2 (DP2, also known as CRTH2) antagonist. Timapiprant (OC000459) potently displaces [3H] PGD2 from human recombinant DP2 (Ki=13 nM), rat recombinant DP2 (Ki=3 nM), and human native DP2 (Ki=4 nM). Timapiprant (OC000459) inhibits mast cell activation of Th2 lymphocytes and eosinophils.

Timapiprant (OC000459) is a potent, selective, and orally active D prostanoid receptor 2 (DP2, also known as CRTH2) antagonist. Timapiprant (OC000459) potently displaces [3H] PGD2 from human recombinant DP2 (Ki=13 nM), rat recombinant DP2 (Ki=3 nM), and human native DP2 (Ki=4 nM). Timapiprant (OC000459) inhibits mast cell activation of Th2 lymphocytes and eosinophils.

Tekniske data

Kemisk navn eller materiale Timapiprant
CAS 851723-84-7
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C21H17FN2O2
Mængde 50 mg
Synonym OC000459
Opløselighedsinformation DMSO : 10 mg/mL (28.71 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL O=C(O)CN1C(C)=C(CC2=NC3=CC=CC=C3C=C2)C4=C1C=CC(F)=C4
Molekylvægt (g/mol) 348.37
Formel vægt 348.37
Procent renhed 98.02%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.