Få mere at vide
Tiglic Acid-d3 (Major), TRC
Artikelnummer. 30452586
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452586 500 mg
30449313 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452586 Leverandør Toronto Research Chemicals Leverandørnr. TRCT440052500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19146-57-7
Molekylær formel C5 2H3 H5 O2
Molekylvægt (g/mol) 103.13
Synonym Crotonic-4,4,4-d3 acid, 2-methyl-, (E)- (8CI), Tiglic acid-d3, (2E)-2-Methyl-2-butenoic-4,4,4-d3 acid, trans-2-Methyl-3-(methyl-d3)acrylic acid, (E)-2-Methyl-2-butenoic acid (4,4,4-D3), Tiglic acid (4,4,4-D3)
IUPAC navn (E)-2-(trideuteriomethyl)but-2-enoic acid
SMIL [2H]C([2H])([2H])\C(=C/C)\C(=O)O
Tekniske data
Kemisk navn eller materiale Tiglic Acid-d3
Mængde 500 mg
InChI formel InChI=1S/C5H8O2/c1-3-4(2)5(6)7/h3H,1-2H3,(H,6,7)/b4-3+/i2D3
Formel vægt 103.0713