Få mere at vide
Thermo Scientific™ Ticlopidine Hydrochloride
Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines
503.89 DKK - 1297.20 DKK
Kemiske identifikatorer
| CAS | 53885-35-1 |
|---|---|
| Formel vægt | 300.25g/mol |
| Synonym | 5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride |
| Format | Powder |
| Til brug med (applikation) | Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines |
| Oversigt | White |
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| 53885-35-1 | |
| 5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride | |
| Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines |
| 300.25g/mol | |
| Powder | |
| White |
Tekniske data
| White | |
| Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines | |
| C14H14ClNS·HCl | |
| 5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride |
| Powder | |
| 53885-35-1 | |
| 300.25g/mol |
Sikkerhed og håndtering
missing translation for 'einecsNumber' : 258-837-4
Anbefalet opbevaring : Ambient temperatures
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.