Få mere at vide
Thiodiphosphoric Acid Tetramethyl Ester (>70%), TRC
Artikelnummer. 30456544
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456544 1 g
30453039 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30456544 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3503501G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51120-35-5
Molekylær formel C4H12O5P2S2
Molekylvægt (g/mol) 266.21
Synonym Thiodiphosphoric Acid ([(HO)2P(S)]2O) Tetramethyl Ester, Destetramethyl Sulfotep
IUPAC navn dimethoxyphosphinothioyloxy-dimethoxy-sulfanylidene-λ^{5}-phosphane
SMIL COP(=S)(OC)OP(=S)(OC)OC
Tekniske data
Kemisk navn eller materiale Thiodiphosphoric Acid Tetramethyl Ester
Mængde 1 g
InChI formel InChI=1S/C4H12O5P2S2/c1-5-10(12,6-2)9-11(13,7-3)8-4/h1-4H3
Formel vægt 265.9601