Få mere at vide
Thiabendazole, 98+%, Thermo Scientific™
Fungicide and antihelminthic
310.65 DKK - 4709.03 DKK
Kemiske identifikatorer
| CAS | 148-79-8 |
|---|---|
| Molekylær formel | C10H7N3S |
| Molekylvægt (g/mol) | 201.247 |
| MDL nummer | MFCD00005587 |
| InChI nøgle | WJCNZQLZVWNLKY-UHFFFAOYSA-N |
| Synonym | thiabendazole, tiabendazole, mintezol, equizole, tiabendazol, mintesol, omnizole, thibenzole, thiabendazol, bovizole |
| PubChem CID | 5430 |
| ChEBI | CHEBI:45979 |
| IUPAC navn | 4-(1H-benzimidazol-2-yl)-1,3-thiazole |
| SMIL | C1=CC=C2C(=C1)NC(=N2)C3=CSC=N3 |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
15402859
|
Thermo Scientific Alfa Aesar
J60009.09 |
10g |
344.78 DKK 310.65 DKK / 10g Spare 34.13 DKK 9% Slukket |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15412859
|
Thermo Scientific Alfa Aesar
J60009.22 |
100g |
1380.62 DKK 1242.65 DKK / 100g Spare 137.97 DKK 9% Slukket |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15422859
|
Thermo Scientific Alfa Aesar
J60009.36 |
500g |
4709.03 DKK
500g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in methanol and dimethyl sulfoxide.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Kemiske identifikatorer
| C10H7N3S | |
| MFCD00005587 | |
| thiabendazole, tiabendazole, mintezol, equizole, tiabendazol, mintesol, omnizole, thibenzole, thiabendazol, bovizole | |
| CHEBI:45979 | |
| C1=CC=C2C(=C1)NC(=N2)C3=CSC=N3 |
Tekniske data
| Thiabendazole | |
| C10H7N3S | |
| UN3077 | |
| 14,9289 | |
| Soluble in methanol and dimethyl sulfoxide. | |
| C1=CC=C2C(=C1)NC(=N2)C3=CSC=N3 | |
| 201.247 | |
| CHEBI:45979 | |
| ≥98% |
| 148-79-8 | |
| MFCD00005587 | |
| 611403 | |
| thiabendazole, tiabendazole, mintezol, equizole, tiabendazol, mintesol, omnizole, thibenzole, thiabendazol, bovizole | |
| WJCNZQLZVWNLKY-UHFFFAOYSA-N | |
| 4-(1H-benzimidazol-2-yl)-1,3-thiazole | |
| 5430 | |
| 201.25 |
Sécurité et traitement
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 205-725-8
missing translation for 'rtecsNumber' : DE0700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Thiabendazole
Mention d'avertissement
- Advarsel
Catégorie de danger
- LANGTIDSFARE FOR VANDMILJØET Kronisk kategori 1
- AKUT FARE FOR VANDMILJØET Akut kategori 1
Mention de danger
- H400-Meget giftig for vandlevende organismer.
- H410-Meget giftig med langvarige virkninger for vandlevende organismer.
Conseils de prudence
- P273-Undgå udledning til miljøet.
- P391-Udslip opsamles.
- P501b-Indholdet/beholderen bortskaffes i henhold til de lokale/regionale/nationale/internationale forskrifter.
Informations supplémentaires
- MIXTURE LIST-Contain: Thiabendazole
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.