Få mere at vide
Thiabendazole, 98+%, Thermo Scientific™
Fungicide and antihelminthic
Brand: Thermo Scientific Alfa Aesar J60009.22
| Mængde | 100g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in methanol and dimethyl sulfoxide.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Kemiske identifikatorer
| C10H7N3S | |
| MFCD00005587 | |
| thiabendazole, tiabendazole, mintezol, equizole, tiabendazol, mintesol, omnizole, thibenzole, thiabendazol, bovizole | |
| CHEBI:45979 | |
| C1=CC=C2C(=C1)NC(=N2)C3=CSC=N3 |
Tekniske data
| Thiabendazole | |
| C10H7N3S | |
| 100g | |
| 611403 | |
| thiabendazole, tiabendazole, mintezol, equizole, tiabendazol, mintesol, omnizole, thibenzole, thiabendazol, bovizole | |
| WJCNZQLZVWNLKY-UHFFFAOYSA-N | |
| 4-(1H-benzimidazol-2-yl)-1,3-thiazole | |
| 5430 | |
| 201.25 |
| 148-79-8 | |
| MFCD00005587 | |
| UN3077 | |
| 14,9289 | |
| Soluble in methanol and dimethyl sulfoxide. | |
| C1=CC=C2C(=C1)NC(=N2)C3=CSC=N3 | |
| 201.247 | |
| CHEBI:45979 | |
| ≥98% |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.