Få mere at vide
TG100-115, TRC
Artikelnummer. 30454389
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30454389 5 mg
30454174 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30454389 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3377955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 677297-51-7
Molekylær formel C18H14N6O2
Molekylvægt (g/mol) 346.34
Synonym TG 100115, 6,7-Bis(3-hydroxyphenyl)pteridine-2,4-diamine, TG100-115
IUPAC navn 3-[2,4-diamino-7-(3-hydroxyphenyl)pteridin-6-yl]phenol
SMIL Nc1nc(N)c2nc(c3cccc(O)c3)c(nc2n1)c4cccc(O)c4
Tekniske data
Kemisk navn eller materiale 3,3'-(2,4-Diamino-6,7-pteridinediyl)bis-phenol
Mængde 5 mg
InChI formel InChI=1S/C18H14N6O2/c19-16-15-17(24-18(20)23-16)22-14(10-4-2-6-12(26)8-10)13(21-15)9-3-1-5-11(25)7-9/h1-8,25-26H,(H4,19,20,22,23,24)
Formel vægt 346.1178