missing translation for 'onlineSavingsMsg'
Få mere at vide

Tetramethylenebis(triphenylphosphonium bromide), 98+%

Artikelnummer. 11422307
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
50 g
250 g
Pakningsstørrelse:
10g
250g
50g
Artikelnummer. Mængde unitSize
11422307 250 g 250g
11402307 10 g 10g
11412307 50 g 50g
3 options
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 11422307

Brand: Thermo Scientific Alfa Aesar A14214.30

Request Bulk or Custom Format

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

CAS: 15546-42-6 | C40H38Br2P2 | 740.5 g/mol

Tetramethylenebis(triphenylphosphonium bromide) is used in the synthesis of squalene. It is also used in the conversion of ?-diketones to cyclohexa-1,3-dienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylenebis(triphenylphosphonium bromide) is used in the synthesis of squalene. It is also used in the conversion of ɑ-diketones to cyclohexa-1,3-dienes.

Notes
Hygroscopic. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 15546-42-6
Molekylær formel C40H38Br2P2
Molekylvægt (g/mol) 740.5
MDL nummer MFCD00031714
InChI nøgle WTFXVDWCEPLDDY-UHFFFAOYSA-L
Synonym tetramethylenebis triphenylphosphonium bromide, 1,4-bis triphenylphosphonio butane dibromide, butane-1,4-diylbis triphenylphosphonium bromide, triphenyl 4-triphenylphosphaniumyl butyl phosphanium dibromide, tetramethylenebis triphenylphosphonium dibromide, triphenyl 4-triphenylphosphaniumylbutyl phosphanium dibromide, phosphonium,1,1'-1,4-butanediyl bis 1,1,1-triphenyl-, bromide 1:2, phosphonium,1,1'-1,4-butanediyl bis 1,1,1-triphenyl-,bromide 1:2
PubChem CID 3084237
IUPAC navn triphenyl(4-triphenylphosphaniumylbutyl)phosphanium;dibromide
SMIL C1=CC=C(C=C1)[P+](CCCC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Br-].[Br-]

Tekniske data

Kemisk navn eller materiale Tetramethylenebis(triphenylphosphonium bromide)
Smeltepunkt 292°C to 293°C
Mængde 250 g
Beilstein 3872522
Følsomhed Hygroscopic
Formel vægt 740.51
Procent renhed ≥98%

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.