Få mere at vide
Tetrahydropalmatine, 98%, Thermo Scientific™
546.28 DKK - 1895.55 DKK
Kemiske identifikatorer
| CAS | 2934-97-6 |
|---|---|
| Molekylær formel | C21H25NO4 |
| Molekylvægt (g/mol) | 355.434 |
| MDL nummer | MFCD00214191 |
| InChI nøgle | AEQDJSLRWYMAQI-UHFFFAOYSA-N |
| Synonym | tetrahydropalmatine, rotundine, tetrahydropalmatin, rotundinum, 2,3,9,10-tetramethoxy-5,8,13,13a-tetrahydro-6h-isoquino 3,2-a isoquinoline, corydalis b, 6h-dibenzo a,g quinolizine, 5,8,13,13a-tetrahydro-2,3,9,10-tetramethoxy, gnf-pf-3943, tetrahydropalmatine hydrochloride, 2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5h-isoquinolino 2,1-b isoquinoline |
| PubChem CID | 5417 |
| IUPAC navn | 2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[2,1-b]isoquinoline |
| SMIL | COC1=C(C2=C(CC3C4=CC(=C(C=C4CCN3C2)OC)OC)C=C1)OC |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
15465589
|
Thermo Scientific Alfa Aesar
J63328.03 |
1g |
546.28 DKK
1g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15475589
|
Thermo Scientific Alfa Aesar
J63328.06 |
1895.55 DKK
5g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | ||||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| C21H25NO4 | |
| MFCD00214191 | |
| tetrahydropalmatine, rotundine, tetrahydropalmatin, rotundinum, 2,3,9,10-tetramethoxy-5,8,13,13a-tetrahydro-6h-isoquino 3,2-a isoquinoline, corydalis b, 6h-dibenzo a,g quinolizine, 5,8,13,13a-tetrahydro-2,3,9,10-tetramethoxy, gnf-pf-3943, tetrahydropalmatine hydrochloride, 2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5h-isoquinolino 2,1-b isoquinoline | |
| 2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[2,1-b]isoquinoline |
Tekniske data
| Tetrahydropalmatine | |
| C21H25NO4 | |
| tetrahydropalmatine, rotundine, tetrahydropalmatin, rotundinum, 2,3,9,10-tetramethoxy-5,8,13,13a-tetrahydro-6h-isoquino 3,2-a isoquinoline, corydalis b, 6h-dibenzo a,g quinolizine, 5,8,13,13a-tetrahydro-2,3,9,10-tetramethoxy, gnf-pf-3943, tetrahydropalmatine hydrochloride, 2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5h-isoquinolino 2,1-b isoquinoline | |
| COC1=C(C2=C(CC3C4=CC(=C(C=C4CCN3C2)OC)OC)C=C1)OC | |
| 355.434 | |
| 355.43 |
| 2934-97-6 | |
| MFCD00214191 | |
| AEQDJSLRWYMAQI-UHFFFAOYSA-N | |
| 2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[2,1-b]isoquinoline | |
| 5417 | |
| 98% |
Sikkerhed og håndtering
missing translation for 'tsca' : No
Anbefalet opbevaring : Ambient temperatures