Få mere at vide
Tetrahydro Cortisone, TRC
Artikelnummer. 17918674
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17918674 50 mg
17947324 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17918674 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29337550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 53-05-4
Molekylær formel C21 H32 O5
Molekylvægt (g/mol) 364.48
Synonym 5beta-Tetrahydrocortisone, NSC 76984, THE, Urocortisone,5beta-Pregnane-3alpha,17alpha,21-triol-11,20-dione
IUPAC navn (3R,5R,8S,9S,10S,13S,14S,17R)-3,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,3,4,5,6,7,8,9,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-11-one
SMIL C[C@]12CC[C@@H](O)C[C@H]1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)CC(=O)[C@H]23
Tekniske data
Kemisk navn eller materiale Tetrahydro Cortisone
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C21H32O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12-15,18,22-23,26H,3-11H2,1-2H3/t12-,13-,14+,15+,18-,19+,20+,21+/m1/s1
Formel vægt 364.225
Procent renhed >95%