Få mere at vide
Tetrabutylammonium Fluoride Hydrate, TRC
Artikelnummer. 30416137
Skift visning
Mængde:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416137 50 g
30416880 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30416137 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29122050G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22206-57-1
Molekylær formel C16H36FN(xH2O)
Molekylvægt (g/mol) 279.48
Synonym N,N,N-Tributyl-1-butanaminium Fluoride Hydrate, Tetra-n-butylammonium Fluoride Hydrate, TBAF Hydrate
IUPAC navn tetrabutylazanium;fluoride;hydrate
SMIL O.[F-].CCCC[N+](CCCC)(CCCC)CCCC
Tekniske data
Kemisk navn eller materiale Tetrabutylammonium Fluoride Hydrate
Mængde 50 g
InChI formel InChI=1S/C16H36N.FH.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;/h5-16H2,1-4H3;1H;1H2/q+1;;/p-1
Formel vægt 279.2937