Learn More
Julabo™ Test Tube Racks for Open Heating Bath Circulator
Brand: Julabo™ 8970321
This item is not returnable.
View return policy
Specifications
Test tube rack | |
ED-5, EH-5, F12-ED, F12-EH, F12-MA open heating bath circulator | |
38 Test Tubes, 12/13 mm dia. | |
Stainless Steel |
Product Content Correction
Product Title
Julabo™ Test Tube Racks for Open Heating Bath Circulator > Stainless Steel; Holds 38 Test tubes, Dia.: 12/13mm
Spot an opportunity for improvement?Share a Content Correction