Learn More
Julabo™ Test Tube Racks for Open Heating Bath Circulator
Brand: Julabo™ 8970380
This item is not returnable.
View return policy
Specifications
Test tube rack | |
SW22, SW23, TW12, TW20, TW8 shaking water bath | |
60 Test Tubes, 16/17 mm dia. | |
Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Julabo™ Test Tube Racks for Open Heating Bath Circulator > Polypropylene; Holds 60 Test tubes, Dia.: 16/17mm
Spot an opportunity for improvement?Share a Content Correction