Få mere at vide
tert-Butyl Methyl-d3 Ether, TRC
Artikelnummer. 30337839
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30337839 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30337839 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69337150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale tert-Butyl Methyl-d3 Ether
CAS 29366-08-3
Molekylær formel C5 D3 H9 O
Mængde 50 mg
Synonym Methyl-tert-Butyl Ether D3, Ether, tert-butyl methyl-d3 (8CI), 2-(Methoxy-d3)-2-methylpropane, Methyl-d3 tert-butyl ether
InChI formel InChI=1S/C5H12O/c1-5(2,3)6-4/h1-4H3/i4D3
SMIL [2H]C([2H])([2H])OC(C)(C)C
IUPAC navn 2-methyl-2-(trideuteriomethoxy)propane
Molekylvægt (g/mol) 91.167
Formel vægt 91.108