Få mere at vide
tert-Butyl Methyl-d3 Ether, TRC
Artikelnummer. 30180421
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180421 100 mg
30180219 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30180421 Leverandør Toronto Research Chemicals Leverandørnr. TRCB693371100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 29366-08-3
Molekylær formel C5 D3 H9 O
Molekylvægt (g/mol) 91.167
Synonym Methyl-tert-Butyl Ether D3, Ether, tert-butyl methyl-d3 (8CI), 2-(Methoxy-d3)-2-methylpropane, Methyl-d3 tert-butyl ether
IUPAC navn 2-methyl-2-(trideuteriomethoxy)propane
SMIL [2H]C([2H])([2H])OC(C)(C)C
Tekniske data
Kemisk navn eller materiale tert-Butyl Methyl-d3 Ether
Mængde 100 mg
InChI formel InChI=1S/C5H12O/c1-5(2,3)6-4/h1-4H3/i4D3
Formel vægt 91.108