Få mere at vide
tert-Butyl Isopropyl Ether, TRC
Artikelnummer. 30134966
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134966 500 mg
30009848 1 g
30009836 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134966 Leverandør Toronto Research Chemicals Leverandørnr. TRCB692450500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17348-59-3
Molekylær formel C7 H16 O
Molekylvægt (g/mol) 116.2
Synonym 2-Methyl-2-(1-methylethoxy)propane, 2-Isopropoxy-2-methylpropane, Isopropyl tert-Butyl Ether;
IUPAC navn 2-methyl-2-propan-2-yloxypropane
SMIL CC(C)OC(C)(C)C
Tekniske data
Kemisk navn eller materiale tert-Butyl Isopropyl Ether
Mængde 500 mg
InChI formel InChI=1S/C7H16O/c1-6(2)8-7(3,4)5/h6H,1-5H3
Formel vægt 116.1201