Få mere at vide
Tenuazonic Acid (>90%), TRC
Artikelnummer. 30010380
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010380 25 mg
18093520 5 mg
17979739 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010380 Leverandør Toronto Research Chemicals Leverandørnr. TRCT01962525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 610-88-8
Molekylær formel C10 H15 N O3
Molekylvægt (g/mol) 197.23
Synonym 2H-Pyrrol-2-one, 3-acetyl-1,5-dihydro-4-hydroxy-5-(1-methylpropyl)-, [S-(R*,R*)]-, 3-Pyrrolin-2-one, 3-acetyl-5-sec-butyl-4-hydroxy-, L- (8CI), Tenuazonic acid (6CI), (5S)-3-Acetyl-1,5-dihydro-4-hydroxy-5-[(1S)-1-methylpropyl]-2H-pyrrol-2-one, AAC-toxin, L-Tenuazonic acid
IUPAC navn (2S)-4-acetyl-2-[(2S)-butan-2-yl]-3-hydroxy-1,2-dihydropyrrol-5-one
SMIL CC[C@H](C)[C@@H]1NC(=O)C(=C1O)C(=O)C
Tekniske data
Kemisk navn eller materiale Tenuazonic acid
Mængde 25 mg
InChI formel InChI=1S/C10H15NO3/c1-4-5(2)8-9(13)7(6(3)12)10(14)11-8/h5,8,13H,4H2,1-3H3,(H,11,14)/t5-,8-/m0/s1
Formel vægt 197.1052