Learn More
Thermo Scientific Chemicals Tenofovir
CAS: 147127-20-6 | C9H14N5O4P, C9 H14 N5 O4 P | 287.22 g/mol
474.00 DKK - 1340.00 DKK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16697032
|
Thermo Scientific Chemicals
465610010 |
1 g |
474.00 DKK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16607042
|
Thermo Scientific Chemicals
465610050 |
5 g |
1340.00 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
147127-20-6 | |
C9 H14 N5 O4 P |
Specifications
147127-20-6 | |
287.21 | |
Reagent | |
SGOIRFVFHAKUTI-ZCFIWIBFSA-N | |
({[(2R)-1-(6-amino-9H-purin-9-yl)propan-2-yl]oxy}methyl)phosphonic acid | |
Tenofovir |
C9 H14 N5 O4 P | |
Powder | |
≥97.5% (HPLC) | |
C[C@H](CN1C=NC2=C(N)N=CN=C12)OCP(O)(O)=O | |
287.22 |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.