Få mere at vide
Telcagepant, MedChemExpress
Artikelnummer. 16483957
Encompass_Preferred
Skift visning
Mængde:
2 mg
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16483957 50 mg -
16490336 2 mg -
16437816 100 mg -
16420508 5 mg 5mg
16488038 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16483957 Leverandør Medchem Express Leverandørnr. HY32709
MedChemExpress Telcagepant (MK-0974) is an orally active calcitonin gene-related peptide (CGRP) receptor antagonist with Kis of 0.77 nM and 1.2 nM for human and rhesus CGRP receptors, respectively.
Telcagepant (MK-0974) is an orally active calcitonin gene-related peptide (CGRP) receptor antagonist with Kis of 0.77 nM and 1.2 nM for human and rhesus CGRP receptors, respectively.
Tekniske data
Kemisk navn eller materiale Telcagepant
CAS 781649-09-0
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C26H27F5N6O3
Mængde 50 mg
Synonym MK-0974
Opløselighedsinformation DMSO : ≥ 50 mg/mL (88.26 mM)
SMIL O=C1NC2=C(N1C3CCN(C(N[C@@H]4CC[C@@H](C5=CC=CC(F)=C5F)CN(CC(F)(F)F)C4=O)=O)CC3)C=CC=N2
Molekylvægt (g/mol) 566.52
Formel vægt 566.52
Procent renhed 99.55%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation