Få mere at vide
Tedizolid, MedChemExpress
Artikelnummer. 16482337
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16482337 50 mg
16466208 10 mg
16408807 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16482337 Leverandør Medchem Express Leverandørnr. HY14855
MedChemExpress Tedizolid (TR 700; Torezolid; DA-7157) is a novel oxazolidinone, acting through inhibition of bacterial protein synthesis by binding to 23S ribosomal RNA (rRNA) of the 50S subunit of the ribosome.
Tedizolid (TR 700
Tekniske data
Kemisk navn eller materiale Tedizolid
CAS 856866-72-3
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C17H15FN6O3
Mængde 50 mg
Synonym TR 700 Torezolid DA-7157
Opløselighedsinformation DMSO : 10 mg/mL (27.00 mM; Need ultrasonic)
SMIL O=C1O[C@@H](CO)CN1C2=CC=C(C3=CC=C(C4=NN(C)N=N4)N=C3)C(F)=C2
Molekylvægt (g/mol) 370.34
Formel vægt 370.34
Procent renhed 99.19%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation