Få mere at vide
Tocris Bioscience™ TCS JNK 6o
GREENER_CHOICE
Artikelnummer. 18024352
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18024352 50 mg
18015542 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18024352 Leverandør Tocris Bioscience™ Leverandørnr. 3222/50
Selective JNK inhibitor
ATP-competitive c-Jun N-terminal kinase (JNK) inhibitor (IC50 values are 2, 4 and 52 nM for JNK1, JNK2 and JNK3 respectively). Displays > 1000 fold selectivity over other kinases, including ERK2 and p38. Inhibits c-Jun phosphorylation (EC50 = 920 nM) and prevents collagen-induced platelet aggregation in vitro.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 894804-07-0
Molekylær formel C18H20N4O4
Tekniske data
Kemisk navn eller materiale TCS JNK 6o
Mængde 50 mg
Formel vægt 356.38
Renhed >98%
Molekylvægt (g/mol) 356.382