CAS RN 894804-07-0
IUPAC Navn:
N-(4-amino-5-cyano-6-ethoxypyridin-2-yl)-2-(2,5-dimethoxyphenyl)acetamide
Synonymer:
N-(4-amino-5-cyano-6-ethoxypyridin-2-yl)-2-(2,5-dimethoxyphenyl)acetamide
Molekylvægt (g/mol):
356.38
Molekylformel:
C18H20N4O4
InChi Key:
KQMPRSZTUSSXND-UHFFFAOYSA-N
SMILES:
CCOC1=C(C#N)C(N)=CC(NC(=O)CC2=C(OC)C=CC(OC)=C2)=N1
1
–
2
af
2
Resultater
1
Tocris Bioscience™ TCS JNK 6o
CAS: 894804-07-0 Molekylær formel: C18H20N4O4
2
JNK Inhibitor VIII, MedChemExpress
MedChemExpress JNK Inhibitor VIII (TCS JNK 6o) is a c-Jun N-terminal kinases (JNK-1, -2, and -3) inhibitor with Ki values of 2 nM, 4 nM, 52 nM, respectively, and has IC50 values of 45 nM and 160 nM for JNK-1 and -2, respectively.