missing translation for 'onlineSavingsMsg'
Learn More

TCERG1L Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

1082.11 DKK - 3059.75 DKK

Specifications

Antigen TCERG1L
Koncentration 800 μg/mL
Applikationer Immunohistochemistry, Immunofluorescence, ELISA
Klassifikation Polyclonal
Konjugeret Unconjugated
View More Specs

Products 2
Product Code Brand Mængde Price Quantity & Availability  
Product Code Brand Mængde Price Quantity & Availability  
30234620
missing translation for 'viewDocuments'
Proteintech
20858-1-AP-150UL
150 μL
3059.75 DKK
150µL
View alternative products
missing translation for 'toSaveUpto' 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
30237387
missing translation for 'viewDocuments'
Proteintech
20858-1-AP-20UL
20 μL
1082.11 DKK
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Description

Description

The TCERG1L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TCERG1L. This antibody recognizes human, mouse antigen. The TCERG1L antibody has been validated for the following applications: IHC, IF-P, ELISA analysis.
Specifications

Specifications

TCERG1L
Immunohistochemistry, Immunofluorescence, ELISA
Unconjugated
Rabbit
Human, Mouse
MGC126584
IgG
Antigen affinity purification
Polyclonal Antibody
800 μg/mL
Polyclonal
Liquid
RUO
256536
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.