Få mere at vide
TCERG1L Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20858-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The TCERG1L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TCERG1L. This antibody recognizes human, mouse antigen. The TCERG1L antibody has been validated for the following applications: IHC, IF-P, ELISA analysis.Tekniske data
| TCERG1L | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 256536 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| 800 μg/mL | |
| MGC126584 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Polyclonal Antibody | |
| IgG |