Learn More
Lenz Laborglasinstrumente™ Tall Shape Weighing Bottles
596.00 DKK - 1665.00 DKK
Specifications
Includes | Interchangeable NS stopper, inscription label |
---|---|
Product Line | Bottle |
Product Type | Weighing Bottle |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
10727141
|
Lenz Laborglasinstrumente™
05565740 |
38 mm |
1445.00 DKK
Pack of 6 |
Estimated Shipment: 09-09-2025
Log in to see stock. |
||||||
11718114
|
Lenz Laborglasinstrumente™
05565750 |
48 mm |
1665.00 DKK
Pack of 6 |
Estimated Shipment: 26-08-2025
Log in to see stock. |
||||||
11836094
|
Lenz Laborglasinstrumente™
05565780 |
75 mm |
596.00 DKK
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
Specifications
Interchangeable NS stopper, inscription label | |
Bottle | |
Weighing Bottle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Tall Shape Weighing Bottles