missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TAK-071, MedChemExpress
Description
TAK-071 is a novel, potent and highly selective muscarinic acetylcholine receptor 1 (M1R) positive allosteric modulator. EC50 of TAK-071 M1R agonist activities is 520 nM.
Specifications
Specifications
| Kemisk navn eller materiale | TAK-071 |
| CAS | 1820812-16-5 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C24H24FN3O3 |
| Mængde | 100 mg |
| Opløselighedsinformation | DMSO : 135 mg/mL (320.32 mM; Need ultrasonic) |
| SMIL | O=C1C2=CC(CC3=CC=C(N4C=CC=N4)C=C3)=C(C)C(F)=C2CN1[C@@H]5[C@H](CCOC5)O |
| Molekylvægt (g/mol) | 421.46 |
| Formel vægt | 421.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?