Få mere at vide
TA-01, MedChemExpress
Artikelnummer. 16472778
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16472778 10 mg 10mg
16465517 5 mg -
16429476 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16472778 Leverandør Medchem Express Leverandørnr. HY100114
MedChemExpress TA-01 is a potent CK1 and p38 MAPK inhibitor, with IC50s of 6.4 nM, 6.8 nM, 6.7 nM for CK1ε, CK1δ and p38 MAPK, respectively. TA-01 acts as a cardiogenic inhibitor.
TA-01 is a potent CK1 and p38 MAPK inhibitor, with IC50s of 6.4 nM, 6.8 nM, 6.7 nM for CK1ε, CK1δ and p38 MAPK, respectively. TA-01 acts as a cardiogenic inhibitor.
Tekniske data
Kemisk navn eller materiale TA-01
CAS 1784751-18-3
Farve White
Renhedsgrad noter Research
Molekylær formel C20H12F3N3
Mængde 10 mg
Opløselighedsinformation DMSO : 50 mg/mL (142.32 mM; Need ultrasonic)
SMIL FC(C=C1)=CC=C1C2=C(C3=CC=NC=C3)NC(C4=C(F)C=CC=C4F)=N2
Molekylvægt (g/mol) 351.32
Formel vægt 351.32
Procent renhed 99.77%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-anti-virus