missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
T-bet/TBX21 Antibody (4B10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
2730.00 DKK - 4025.00 DKK
Tekniske data
| Antigen | T-bet/TBX21 |
|---|---|
| Klon | 4B10 |
| Koncentration | 0.5 mg/mL |
| Fortynding | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Flow (Intracellular) |
| Applikationer | Western Blot, Flow Cytometry, Immunoprecipitation |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18262555
|
Novus Biologicals
NBP1-43298-0.025MG |
0.025 mg |
2730.00 DKK
25µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18282922
|
Novus Biologicals
NBP1-43298-0.1MG |
0.1 mg |
4025.00 DKK
0.10mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Intracellular).Tekniske data
| T-bet/TBX21 | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| 30009 | |
| The immunogen for this antibody was TBet. | |
| Primary | |
| Store at 4C. Do not freeze. |
| 4B10 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Flow (Intracellular) | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rhesus Macaque | |
| T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
| TBX21 | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel