missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
T-bet/TBX21 Antibody (4B10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43298-0.1MG
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Intracellular).
Tekniske data
| T-bet/TBX21 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Flow (Intracellular) | |
| T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 30009 | |
| Store at 4C. Do not freeze. |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| 4B10 | |
| Unconjugated | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| TBX21 | |
| The immunogen for this antibody was TBet. | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rhesus Macaque | |
| IgG1 κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion