Learn More
Poulten Selfe™ Suspended-Level Viscometers
Suitable for transparent liquids. With UKAS certificate of calibration.
Brand:  Poulten Selfe™ 1625/08
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13131053
  • 1460.00 DKK / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
Description
Description
  • Type BS/IP/SL, IP71.Suitable for transparent liquids. Nominal overall length 330mm. Approximate sample size 22mL ­to 40mL. Available without certificate of calibration, please enquire.
  • With UKAS certificate of calibration.
Specifications
Specifications
Suspended-Level Viscometer
UKAS
22 to 40 mL
6.000 to 30.000 cSt
4A