Learn More
Poulten Selfe™ Suspended-Level Viscometers
Suitable for transparent liquids. With UKAS certificate of calibration.
1460.00 DKK
Specifications
Product Type | Suspended-Level Viscometer |
---|---|
Certifications/Compliance | UKAS |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
13181043
|
Poulten Selfe™
1625/03 |
2 |
1460.00 DKK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
13161043
|
Poulten Selfe™
1625/01 |
1 |
1460.00 DKK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
13191043
|
Poulten Selfe™
1625/04 |
2A |
1460.00 DKK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
13121053
|
Poulten Selfe™
1625/07 |
4 |
1460.00 DKK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
13111053
|
Poulten Selfe™
1625/06 |
3A |
1460.00 DKK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
13131053
|
Poulten Selfe™
1625/08 |
4A |
1460.00 DKK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
13101053
|
Poulten Selfe™
1625/05 |
3 |
1460.00 DKK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
13141053
|
Poulten Selfe™
1625/09 |
5 |
1460.00 DKK
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
- Type BS/IP/SL, IP71.Suitable for transparent liquids. Nominal overall length 330mm. Approximate sample size 22mL to 40mL. Available without certificate of calibration, please enquire.
- With UKAS certificate of calibration.
Specifications
Suspended-Level Viscometer | |
UKAS |