missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Supersaturated Sea Water Conductivity Calibrator, YSI™
Brand: YSI Yellow Spring Instrument CRT-970-536B
Quantity | 1 Ea. |
---|
This item is not returnable.
View return policy
Description

Specifications
Yes | |
1g/mL | |
6.5 to 7.5 | |
1 Ea. |
Supersaturated Sea Water Conductivity Calibrator | |
Liquid | |
100°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Supersaturated Sea Water Conductivity Calibrator, YSI™ > Conductivity Range: 10000umho
Spot an opportunity for improvement?Share a Content Correction