missing translation for 'onlineSavingsMsg'
Få mere at vide

Sunset Yellow FCF, TRC

Artikelnummer. 30010455
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010455 5 g
30005294 10 g
30007514 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30010455 Leverandør Toronto Research Chemicals Leverandørnr. TRCS8250005G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2783-94-0
Molekylær formel C16 H10 N2 O7 S2 . 2 Na
Molekylvægt (g/mol) 452.37
Synonym 2-Naphthalenesulfonic acid, 6-hydroxy-5-[2-(4-sulfophenyl)diazenyl]-, sodium salt (1:2), 2-Naphthalenesulfonic acid, 6-hydroxy-5-[(4-sulfophenyl)azo]-, disodium salt (9CI), 6-Hydroxy-5-[(4-sulfophenyl)azo]-2-naphthalenesulfonic acid disodium salt, C.I. Food Yellow 3 (7CI), C.I. Food Yellow 3, disodium salt (8CI), Sunset Yellow FCF (6CI), 1-p-Sulfophenylazo-2-hydroxynaphthalene-6-sulfonate disodium salt, 1351 Yellow, 1899 Yellow, Acid Food Yellow 3, Acid Yellow TRA, Aizen Food Yellow 5, Alabaster No. 3, Atul Sunset Yellow FCF, Bright yellow FCF, C Orange 10, C.I. 15985, CI 15985 yellow, Canacert Sunset Yellow FCF, Certicol Sunset Yellow CFS, Cilefa Orange S, Cogilor Orange 213.11, D and C Yellow 6, D and C Yellow No. 6, D&C Yellow 6, D&C Yellow No. 6, Dolkwal Sunset Yellow, E 110, E 110 (dye), Edicol Supra Yellow FC, Eniacid Sunset Yellow, Eurocert Orange FCF, FD & C Yellow 6, FD 155, FD and C Yellow 6, FD and C Yellow No. 6, FD&C Yellow No.6, Food Orange, Food Yellow 3, Food Yellow 5, Food Yellow No. 5, HD Sunset Yellow FCF, HD Sunset Yellow FCF Supra, Hexacol Sunset Yellow FCF, Hexacol Sunset Yellow FCF Supra, INS 110, Japan Food Yellow No. 5, Japan Yellow 5, Japan Yellow No. 5, KCA Foodcol Sunset Yellow FCF, L Orange Z 2010, Maple Sunset Yellow FCF, Orange II R, Orange PAL, Orange Yellow 85, Orange Yellow S, Orange Yellow SFQ, Orient Water Yellow 2, Para Orange, Sun Orange A Geigy, Sun Yellow, Sun Yellow Extra Conc. A Export, Sun Yellow Extra Pure A, Sun Yellow FCF, Sunlight Yellow FCF, Sunset Yellow, Sunset Yellow 6, Sunset Yellow E 110, Twilight yellow, Usacert Yellow No. 6, Vitasyn Orange RGL 90, Water Yellow 2, Yellow 6, Yellow No. 5, Yellow No. 6, Yellow Orange S, Yellow S, Yellow SY, Yellow Sun
IUPAC navn disodium;6-hydroxy-5-[(E)-(4-sulfonatophenyl)diazenyl]naphthalene-2-sulfonate
SMIL [Na+].[Na+].Oc1ccc2cc(ccc2c1N=Nc3ccc(cc3)S(=O)(=O)[O-])S(=O)(=O)[O-]

Tekniske data

Kemisk navn eller materiale Sunset Yellow (E110)
Renhedsgrad noter HPLC
Mængde 5 g
InChI formel InChI=1S/C16H12N2O7S2.2Na/c19-15-8-1-10-9-13(27(23,24)25)6-7-14(10)16(15)18-17-11-2-4-12(5-3-11)26(20,21)22;;/h1-9,19H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2
Formel vægt 451.9725
Procent renhed >95%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.